Gene Transcripts

Gene Symbol
Wap
Entrez Gene ID
114596
Gene Name
whey acidic protein
Gene Aliases
Chromosome Location
Chr. 14: 86897948 - 86900412 on Build Rnor_6.0
UniGene ID
Rn.9978
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053751.4 NP_446203.1 3 322
XM_008770244.1 XP_008768466.1 3 689
GenBank mRNA J00801.1 - 3 319
J00802.1 - 3 322

View Full Product Details