Gene Transcripts

Gene Symbol
Cry2
Entrez Gene ID
170917
Gene Name
cryptochrome circadian clock 2
Gene Aliases
Chromosome Location
Chr. 3: 81314151 - 81344109 on Build Rnor_6.0
UniGene ID
Rn.21150
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133405.2 NP_596896.2 3 348
GenBank mRNA AY033877.1 - 3 358
BC166416.1 - 3 348 9033484
FQ211347.1 - 3 358

View Full Product Details