Gene Transcripts

Gene Symbol
Jmjd1c
Entrez Gene ID
171120
Gene Name
jumonji domain containing 1C
Gene Aliases
Pr2
Chromosome Location
Chr. 20: 22751743 - 22914108 on Build Rnor_6.0
UniGene ID
Rn.99017
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191719.1 NP_001178648.1 8 1884
XM_006256359.2 XP_006256421.1 8 1935
XM_006256360.2 XP_006256422.1 7 1731
XM_006256361.2 XP_006256423.1 8 1958
XM_006256362.2 XP_006256424.1 8 1850
XM_006256363.1 XP_006256425.1 4 1292
XM_006256364.1 XP_006256426.1 8 1844
XM_006256365.1 XP_006256427.1 3 1150

View Full Product Details