Gene Transcripts

Gene Symbol
Vom2r27
Entrez Gene ID
286914
Gene Name
vomeronasal 2 receptor, 27
Gene Aliases
Chromosome Location
Chr. 1: 64667030 - 64699156 on Build Rnor_6.0
UniGene ID
Rn.10810
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173130.1 NP_775153.1 4 1395
XM_006228015.1 XP_006228077.1 4 1397
GenBank mRNA AF016182.1 - 4 1395

View Full Product Details