Gene Transcripts

Gene Symbol
TRAPPC10
Entrez Gene ID
7109
Gene Name
trafficking protein particle complex 10
Gene Aliases
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
Chromosome Location
Chr. 21: 44012325 - 44106552 on Build GRCh38
UniGene ID
Hs.126221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003274.4 NP_003265.3 4 545
XM_005261168.4 XP_005261225.1 4 538
XM_011529714.2 XP_011528016.1 4 538
XM_011529715.2 XP_011528017.1 4 538
XM_011529716.2 XP_011528018.1 4 538
XM_011529717.2 XP_011528019.1 4 538
XM_011529718.2 XP_011528020.1 4 538
XM_011529719.2 XP_011528021.1 4 538
XM_011529720.2 XP_011528022.1 4 538
XM_011529721.2 XP_011528023.1 1 24
XM_011529725.2 XP_011528027.1 4 538
XM_011529726.2 XP_011528028.1 4 538
XM_011529728.2 XP_011528030.1 1 55
XM_011529730.2 XP_011528032.1 4 538
XM_017028454.1 XP_016883943.1 4 538
GenBank mRNA AL559301.3 - 4 551
BC046241.1 - 4 523
BC052247.1 - 4 523
BC094823.1 - 4 523
BC101728.1 - 4 465 8069234
BF979287.1 - 4 563
BX370317.2 - 1 27
U19252.1 - 4 513
U61500.1 - 4 529

View Full Product Details