Gene Transcripts

Gene Symbol
Pigx
Entrez Gene ID
288041
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class X
Gene Aliases
RGD1307289
Chromosome Location
Chr. 11: 71938388 - 71954225 on Build Rnor_6.0
UniGene ID
Rn.34037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100651.1 NP_001094121.1 3, 4 467
GenBank mRNA AB177393.1 - 3, 4 467
DY311520.1 - 3, 4 465
FQ229771.1 - 500

View Full Product Details