Gene Transcripts

Gene Symbol
Vom2r30
Entrez Gene ID
289295
Gene Name
vomeronasal 2 receptor, 30
Gene Aliases
RGD1561133
Chromosome Location
Chr. 1: 74770009 - 74791415 on Build Rnor_6.0
UniGene ID
Rn.217650
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099467.1 NP_001092937.1 3 1063

View Full Product Details