Gene Transcripts

Gene Symbol
Dleu7
Entrez Gene ID
290308
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
RGD1305071
Chromosome Location
Chr. 15: 45360503 - 45376807 on Build Rnor_6.0
UniGene ID
Rn.45118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252176.2 XP_006252238.1 1, 2 1034
GenBank mRNA BC158600.1 - 1, 2 551

View Full Product Details