Gene Transcripts

Gene Symbol
Olfm4
Entrez Gene ID
290409
Gene Name
olfactomedin 4
Gene Aliases
Chromosome Location
Chr. 15: 62384262 - 62429450 on Build Rnor_6.0
UniGene ID
Rn.156265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106052.1 NP_001099522.1 5 957
XM_008770863.1 XP_008769085.1 5 1784

View Full Product Details