Gene Transcripts

Gene Symbol
Slc35d2
Entrez Gene ID
290959
Gene Name
solute carrier family 35 (UDP-GlcNAc/UDP-glucose transporter), member D2
Gene Aliases
Chromosome Location
Chr. 17: 1618641 - 1649983 on Build Rnor_6.0
UniGene ID
Rn.144756
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106098.2 NP_001099568.2 7 574
XM_006253493.2 XP_006253555.1 7 1558

View Full Product Details