Gene Transcripts

Gene Symbol
Mcoln2
Entrez Gene ID
292168
Gene Name
mucolipin 2
Gene Aliases
Chromosome Location
Chr. 2: 252018594 - 252068209 on Build Rnor_6.0
UniGene ID
Rn.35113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039005.1 NP_001034094.1 6, 7 789
GenBank mRNA BC099787.1 - 6, 7 789

View Full Product Details