Gene Transcripts

Gene Symbol
Rnaset2
Entrez Gene ID
292306
Gene Name
ribonuclease T2
Gene Aliases
Chromosome Location
Chr. 1: 53174879 - 53192048 on Build Rnor_6.0
UniGene ID
Rn.18019
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106210.1 NP_001099680.1 6, 7 554
GenBank mRNA BC168957.1 - 587

View Full Product Details