Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
293180
Gene Name
MICAL C-terminal like
Gene Aliases
RSB-11-77
Chromosome Location
Chr. 1: 177193735 - 177232674 on Build Rnor_6.0
UniGene ID
Rn.30134
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182669.2 NP_872610.2 4 575
GenBank mRNA BC098647.1 - 4 575
BC100137.1 - 1 26 7457561

View Full Product Details