Gene Transcripts

Gene Symbol
USP1
Entrez Gene ID
7398
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
UBP
Chromosome Location
Chr. 1: 62436304 - 62451804 on Build GRCh38
UniGene ID
Hs.35086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017415.1 NP_001017415.1 6 903
NM_001017416.1 NP_001017416.1 6 808
NM_003368.4 NP_003359.3 6 1333
GenBank mRNA AB014458.1 - 6 684
AB208893.1 - 6 965
AF117386.1 - 6 820
AK312423.1 - 6 996
AL117575.1 - 5 655
BC018745.1 - 6 1333
BC023216.1 - 2 168
BC032364.1 - 6 983
BC050525.1 - 6 1037 6473568

View Full Product Details