Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007126.3 | NP_009057.1 | 14 | 2149 | |
GenBank mRNA | AF100752.1 | - | 14 | 1972 | |
AK307735.1 | - | 14 | 1964 | ||
AK312310.1 | - | 14 | 2031 | ||
AL137377.1 | - | 5 | 635 | ||
BC007562.1 | - | 10 | 1275 | ||
BC110913.1 | - | 14 | 1959 | 6502535 | |
BC121794.1 | - | 14 | 1956 | 40108360 | |
BC122550.1 | - | 13 | 2034 | 40108357 | |
FJ224344.1 | - | 14 | 1972 | ||
FJ224352.1 | - | 14 | 1972 |