Gene Transcripts

Gene Symbol
Brsk2
Entrez Gene ID
293631
Gene Name
BR serine/threonine kinase 2
Gene Aliases
Chromosome Location
Chr. 1: 214924245 - 214949643 on Build Rnor_6.0
UniGene ID
Rn.232994
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760049.1 XP_008758271.1 2 160
GenBank mRNA AM040976.1 - 412

View Full Product Details