Gene Transcripts

Gene Symbol
Cstl1
Entrez Gene ID
296220
Gene Name
cystatin-like 1
Gene Aliases
Chromosome Location
Chr. 3: 143084557 - 143089554 on Build Rnor_6.0
UniGene ID
Rn.27784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106522.1 NP_001099992.1 3, 4 502

View Full Product Details