Gene Transcripts

Gene Symbol
Abo
Entrez Gene ID
100301568
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Gene Aliases
Abo2
Chromosome Location
Chr. 3: 4374679 - 4394374 on Build Rnor_6.0
UniGene ID
Rn.112591
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160263.1 NP_001153735.1 3, 4 184
GenBank mRNA AB081652.1 - 3, 4 228
AY228143.1 - 3, 4 184

View Full Product Details