Gene Transcripts

Gene Symbol
C8g
Entrez Gene ID
296545
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
Chromosome Location
Chr. 3: 2724616 - 2727616 on Build Rnor_6.0
UniGene ID
Rn.17399
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106555.1 NP_001100025.1 4 496
XM_006233581.2 XP_006233643.1 4 881
GenBank mRNA FQ210613.1 - 4 456
FQ218819.1 - 4 456

View Full Product Details