Gene Transcripts

Gene Symbol
Lcn9
Entrez Gene ID
296578
Gene Name
lipocalin 9
Gene Aliases
RGD1306341
Chromosome Location
Chr. 3: 3272173 - 3274715 on Build Rnor_6.0
UniGene ID
Rn.218664
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106560.1 NP_001100030.1 2 204
XM_008761622.1 XP_008759844.1 2 224
GenBank mRNA DQ537496.1 - 2 193

View Full Product Details