Gene Transcripts

Gene Symbol
Dph2
Entrez Gene ID
298452
Gene Name
DPH2 homolog (S. cerevisiae)
Gene Aliases
Dph2l2
Chromosome Location
Chr. 5: 136723398 - 136726358 on Build Rnor_6.0
UniGene ID
Rn.49365
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015007.1 NP_001015007.1 4 1162
GenBank mRNA BC092656.1 - 4 1162

View Full Product Details