Gene Transcripts

Gene Symbol
Smap2
Entrez Gene ID
298500
Gene Name
small ArfGAP2
Gene Aliases
RGD1308418, Smap1l
Chromosome Location
Chr. 5: 139885321 - 139931366 on Build Rnor_6.0
UniGene ID
Rn.203765
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100669.1 NP_001094139.1 3 665
GenBank mRNA BC161927.1 - 3 659

View Full Product Details