Gene Transcripts

Gene Symbol
Agmat
Entrez Gene ID
298607
Gene Name
agmatine ureohydrolase (agmatinase)
Gene Aliases
Chromosome Location
Chr. 5: 160306759 - 160322127 on Build Rnor_6.0
UniGene ID
Rn.7880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048185.1 NP_001041650.1 4 677
XM_008764233.1 XP_008762455.1 4 869
GenBank mRNA BC091231.1 - 4 661
BC105628.1 - 4 677 7322772

View Full Product Details