Gene Transcripts

Gene Symbol
Ylpm1
Entrez Gene ID
299199
Gene Name
YLP motif containing 1
Gene Aliases
RGD1564946, Zap, Zap3
Chromosome Location
Chr. 6: 108820774 - 108897209 on Build Rnor_6.0
UniGene ID
Rn.41317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271258.1 NP_001258187.1 9 5276

View Full Product Details