Gene Transcripts

Gene Symbol
Acyp1
Entrez Gene ID
299203
Gene Name
acylphosphatase 1, erythrocyte (common) type
Gene Aliases
Chromosome Location
Chr. 6: 109097065 - 109104058 on Build Rnor_6.0
UniGene ID
Rn.3465
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106746.2 NP_001100216.1 2, 3 250
NM_001199167.1 NP_001186096.1 2, 3 182
NM_001199168.1 NP_001186097.1 2, 3 146
XM_006240308.2 XP_006240370.1 2, 3 301
GenBank mRNA FM034312.1 - 2, 3 146
FM062949.1 - 2, 3 238

View Full Product Details