Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
299314
Gene Name
complement factor properdin
Gene Aliases
Pfc
Chromosome Location
Chr. X: 1311121 - 1316683 on Build Rnor_6.0
UniGene ID
Rn.204119
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106757.2 NP_001100227.1 7 995
GenBank mRNA BC158889.1 - 7 995
FQ209917.1 - 7 1028
FQ212442.1 - 7 1030

View Full Product Details