Gene Transcripts

Gene Symbol
Slc25a47
Entrez Gene ID
299316
Gene Name
solute carrier family 25, member 47
Gene Aliases
Hdmcp, Slc24a47
Chromosome Location
Chr. 6: 132762306 - 132767940 on Build Rnor_6.0
UniGene ID
Rn.109623
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001509.1 NP_001001509.1 2, 3 111
GenBank mRNA AY603424.1 - 2, 3 111
BC089874.1 - 2, 3 115 7325453
FQ210765.1 - 2, 3 171

View Full Product Details