Gene Transcripts

Gene Symbol
Ndufb9
Entrez Gene ID
299954
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9
Gene Aliases
Chromosome Location
Chr. 7: 98813062 - 98819408 on Build Rnor_6.0
UniGene ID
Rn.22045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127294.2 NP_001120766.1 2 162
GenBank mRNA BC166927.1 - 2 162
BC168767.1 - 2 162 6891022
FQ210477.1 - 2 169
FQ217145.1 - 2 168
FQ217579.1 - 2 181
FQ217728.1 - 2 162
FQ220460.1 - 2 181
FQ223542.1 - 2 169
FQ223611.1 - 2 181
FQ224000.1 - 2 227
FQ224351.1 - 2 181
FQ224618.1 - 2 198
FQ224680.1 - 2 169
FQ228493.1 - 2 169
FQ228673.1 - 2 181

View Full Product Details