Gene Transcripts

Gene Symbol
Zc3h3
Entrez Gene ID
300032
Gene Name
zinc finger CCCH type containing 3
Gene Aliases
Zc3hdc3
Chromosome Location
Chr. 7: 116787762 - 116872558 on Build Rnor_6.0
UniGene ID
Rn.198237
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134865.1 NP_001128337.1 4 1733
XM_008765548.1 XP_008763770.1 4 1755

View Full Product Details