Gene Transcripts

Gene Symbol
Ldoc1l
Entrez Gene ID
300114
Gene Name
leucine zipper, down-regulated in cancer 1-like
Gene Aliases
RGD1306472
Chromosome Location
Chr. 7: 125461408 - 125466517 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242147.2 XP_006242209.1 1 1034

View Full Product Details