Gene Transcripts

Gene Symbol
Or10ad1
Entrez Gene ID
500926
Gene Name
olfactory receptor, family 10, subfamily AD, member 1
Gene Aliases
RGD1562157
Chromosome Location
Chr. 7: 139781775 - 139782794 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765883.1 XP_008764105.1 1 559

Gene Symbol
Olr1102
Entrez Gene ID
300194
Gene Name
olfactory receptor 1102
Gene Aliases
Or10ad1, ratchr7-137003272-137002316_ORF
Chromosome Location
Chr. 7: 139802539 - 139805179 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765851.1 XP_008764073.1 2 551

View Full Product Details