Gene Transcripts

Gene Symbol
Aasdhppt
Entrez Gene ID
300328
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
Chromosome Location
Chr. 8: 1439330 - 1450095 on Build Rnor_6.0
UniGene ID
Rn.17217
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106798.2 NP_001100268.2 2 321
GenBank mRNA BC098729.1 - 1 677
BC166800.1 - 2 321 7938259
FQ209549.1 - 2 337
FQ214028.1 - 2 340
FQ214274.1 - 2 334
FQ214451.1 - 2 335
FQ220704.1 - 2 335
FQ230080.1 - 2 340

View Full Product Details