Gene Transcripts

Gene Symbol
Ttc21a
Entrez Gene ID
301065
Gene Name
tetratricopeptide repeat domain 21A
Gene Aliases
RGD1308806, Ttc21a-ps1
Chromosome Location
Chr. 8: 128622403 - 128657199 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750604.3 XP_003750652.1 8 929
XM_006244150.2 XP_006244212.1 8 929

View Full Product Details