Gene Transcripts

Gene Symbol
Guca1a
Entrez Gene ID
301233
Gene Name
guanylate cyclase activator 1a (retina)
Gene Aliases
Chromosome Location
Chr. 9: 15609804 - 15620030 on Build Rnor_6.0
UniGene ID
Rn.39165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106887.1 NP_001100357.1 2 346
XM_006244428.2 XP_006244490.1 1 301

View Full Product Details