Gene Transcripts

Gene Symbol
Dach2
Entrez Gene ID
302333
Gene Name
dachshund family transcription factor 2
Gene Aliases
RGD1561687
Chromosome Location
Chr. X: 85086864 - 85484150 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257198.2 XP_006257260.1 4 1081

View Full Product Details