Gene Transcripts

Gene Symbol
Srl
Entrez Gene ID
302948
Gene Name
sarcalumenin
Gene Aliases
Chromosome Location
Chr. 10: 11240141 - 11284325 on Build Rnor_6.0
UniGene ID
Rn.233843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750771.3 XP_003750819.1 4 1551
XM_008767510.1 XP_008765732.1 3 257

View Full Product Details