Gene Transcripts

Gene Symbol
Atad5
Entrez Gene ID
303348
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
RGD1561464
Chromosome Location
Chr. 10: 67427042 - 67474838 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246963.2 XP_006247025.1 19 4614
XM_220750.8 XP_220750.6 20 4803

View Full Product Details