Gene Transcripts

Gene Symbol
Ttc25
Entrez Gene ID
303534
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
RGD1311859
Chromosome Location
Chr. 10: 88458636 - 88488237 on Build Rnor_6.0
UniGene ID
Rn.12620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107049.1 NP_001100519.1 9 1380
XM_006247310.2 XP_006247372.1 9 2234
GenBank mRNA BC162060.1 - 9 1323

View Full Product Details