Gene Transcripts

Gene Symbol
Wipi1
Entrez Gene ID
303630
Gene Name
WD repeat domain, phosphoinositide interacting 1
Gene Aliases
RGD1307754
Chromosome Location
Chr. 10: 97859730 - 97896886 on Build Rnor_6.0
UniGene ID
Rn.203725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127297.1 NP_001120769.1 5 547
XM_008768366.1 XP_008766588.1 5 1166
XM_008768367.1 XP_008766589.1 5 444
GenBank mRNA BC166829.1 - 5 547

View Full Product Details