Gene Transcripts

Gene Symbol
Cpn2
Entrez Gene ID
303861
Gene Name
carboxypeptidase N, polypeptide 2
Gene Aliases
RGD1305170
Chromosome Location
Chr. 11: 74057374 - 74067486 on Build Rnor_6.0
UniGene ID
Rn.126621
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107085.2 NP_001100555.1 2 451
XM_006248485.2 XP_006248547.1 3 508
GenBank mRNA BC160859.1 - 1 387

View Full Product Details