Gene Transcripts

Gene Symbol
Sbno1
Entrez Gene ID
304470
Gene Name
strawberry notch homolog 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 12: 37593692 - 37653772 on Build Rnor_6.0
UniGene ID
Rn.12294
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107138.1 NP_001100608.1 17 2300
XM_006249300.2 XP_006249362.1 17 2877
XM_006249301.2 XP_006249363.1 17 2874
XM_008769224.1 XP_008767446.1 16 2772
XM_008769225.1 XP_008767447.1 16 2769
XM_008769226.1 XP_008767448.1 14 1782
GenBank mRNA FQ230302.1 - 8 989
FQ232681.1 - 8 989

View Full Product Details