Gene Transcripts

Gene Symbol
Brox
Entrez Gene ID
305031
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
RGD1307161
Chromosome Location
Chr. 13: 101677896 - 101697738 on Build Rnor_6.0
UniGene ID
Rn.3660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025669.1 NP_001020840.1 3 254
GenBank mRNA BC097348.1 - 3 254

View Full Product Details