Gene Transcripts

Gene Symbol
Cenpu
Entrez Gene ID
306464
Gene Name
centromere protein U
Gene Aliases
Mlf1ip
Chromosome Location
Chr. 16: 48898032 - 48921248 on Build Rnor_6.0
UniGene ID
Rn.128609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025673.1 NP_001020844.1 11, 12 1183
GenBank mRNA BC097399.1 - 11, 12 1183

View Full Product Details