Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006768.4 | NP_006759.3 | 8 | 1218 | |
XM_005253944.4 | XP_005254001.1 | 8 | 1318 | ||
XM_011538789.2 | XP_011537091.1 | 7 | 988 | ||
XM_017019992.1 | XP_016875481.1 | 7 | 1026 | ||
XM_017019993.1 | XP_016875482.1 | 7 | 1009 | ||
XM_017019994.1 | XP_016875483.1 | 6 | 884 | ||
XM_017019995.1 | XP_016875484.1 | 6 | 649 | ||
GenBank mRNA | AB208878.1 | - | 8 | 1173 | |
AF035620.1 | - | 8 | 1153 | ||
AK299326.1 | - | 5 | 524 | ||
AK308279.1 | - | 8 | 1065 | ||
AY332222.1 | - | 8 | 1153 | ||
BC136698.1 | - | 8 | 1153 | 9020688 | |
BC136699.1 | - | 8 | 1153 | 9020689 |