Gene Transcripts

Gene Symbol
Lama3
Entrez Gene ID
307582
Gene Name
laminin, alpha 3
Gene Aliases
Chromosome Location
Chr. 18: 3704864 - 3941170 on Build Rnor_6.0
UniGene ID
Rn.10597
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772008.1 XP_008770230.1 64 9405
GenBank mRNA U61261.1 - 26 3761

View Full Product Details