Gene Transcripts
- Gene Symbol
- Klk14
- Entrez Gene ID
- 308562
- Gene Name
- kallikrein related-peptidase 14
- Gene Aliases
- Chromosome Location
- Chr. 1: 99616183 - 99621425 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_218641.8 |
XP_218641.5
|
2 |
68 |
|
View Full Product Details