Gene Transcripts

Gene Symbol
Klk12
Entrez Gene ID
308564
Gene Name
kallikrein related-peptidase 12
Gene Aliases
Chromosome Location
Chr. 1: 99706148 - 99711004 on Build Rnor_6.0
UniGene ID
Rn.83473
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107508.1 NP_001100978.1 4 665
XM_008759351.1 XP_008757573.1 6 745

View Full Product Details