Gene Transcripts

Gene Symbol
Olr37
Entrez Gene ID
308853
Gene Name
olfactory receptor 37
Gene Aliases
Chromosome Location
Chr. 1: 164755094 - 164756059 on Build Rnor_6.0
UniGene ID
Rn.110414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000504.1 NP_001000504.1 1 47

Gene Symbol
Olr36
Entrez Gene ID
405257
Gene Name
olfactory receptor 36
Gene Aliases
Chromosome Location
Chr. 1: 164717029 - 164717937 on Build Rnor_6.0
UniGene ID
Rn.141937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000935.1 NP_001000935.1 1 47

View Full Product Details