Gene Transcripts

Gene Symbol
Fchsd2
Entrez Gene ID
308864
Gene Name
FCH and double SH3 domains 2
Gene Aliases
Chromosome Location
Chr. 1: 166140117 - 166395915 on Build Rnor_6.0
UniGene ID
Rn.2386
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107539.1 NP_001101009.1 14, 15 1490
XM_008759720.1 XP_008757942.1 16, 17 1754
XM_008759721.1 XP_008757943.1 16, 17 1754
XM_008759722.1 XP_008757944.1 15, 16 1682

View Full Product Details