Gene Transcripts
- Gene Symbol
- ARHGAP6
- Entrez Gene ID
- 395
- Gene Name
- Rho GTPase activating protein 6
- Gene Aliases
- RHOGAP6, RHOGAPX-1
- Chromosome Location
- Chr. X: 11137543 - 11665701 on Build GRCh38
- UniGene ID
- Hs.435291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287242.1 |
NP_001274171.1
|
6, 7 |
977 |
|
NM_006125.2 |
NP_006116.2
|
6, 7 |
2200 |
|
NM_013423.2 |
NP_038267.1
|
6, 7 |
1029 |
|
NM_013427.2 |
NP_038286.2
|
6, 7 |
2200 |
|
NR_109776.1 |
|
7, 8 |
2296 |
|
GenBank mRNA |
AB208792.1 |
- |
6, 7 |
1582 |
|
AF012272.2 |
- |
6, 7 |
2201 |
|
AF022212.2 |
- |
7, 8 |
2297 |
|
AF117067.1 |
- |
6, 7 |
2201 |
|
AF177663.1 |
- |
6, 7 |
1029 |
|
AF177665.1 |
- |
6, 7 |
835 |
|
AK293078.1 |
- |
6, 7 |
1631 |
|
AK297713.1 |
- |
6, 7 |
874 |
|
AK303444.1 |
- |
6, 7 |
890 |
|
BC150635.1 |
- |
6, 7 |
1359 |
9057005 |
View Full Product Details